Order Entry
Germany
Orders LinkContactUsLinkComponent
Human Complement Factor D ELISA Kit

Human Complement Factor D ELISA Kit

Human Complement Factor D ELISA kit, 96 tests
Catalog # ABCAAB243686-1X96
Supplier:   Abcam
Human Complement Factor D ELISA Kit
Catalog # ABCAAB243686-1X96
Supplier:  Abcam
CAS Number:  
Human Complement Factor D ELISA Kit
Human Complement Factor D ELISA Kit

Human Complement Factor D ELISA kit, 96 tests

CAS Number:  
Price
764,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Human Complement Factor D ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Complement Factor D with a sensitivity of 2,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.

Specifications

Specifications

  • Description:
    Human Complement Factor D ELISA kit
  • Tests per kit:
    96 tests
  • Primary antibody reactivity:
    Human
  • Sample type:
    Urine, heparin plasma, citrate plasma, cell culture supernatant, serum, EDTA plasma
  • Detection range:
    15,63 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB243686-1X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    2,6 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Factor D/CFD
  • Regulatory status:
    RUO

Specifications