Mouse E-Selectin ELISA Kit (CD62E)
Mouse E-Selectin ELISA kit (CD62E), 96 tests
Catalog # ABCAAB201279-96
Supplier: Abcam
Jump to:
Product Details & Documents
Mouse E-Selectin ELISA kit (CD62E) is a single-wash 90-min Simplestep used to quantify Mouse E-Selectin (CD62E) with a sensitivity of 8.55 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.
Specifications
Specifications
- Description:Mouse E-Selectin ELISA kit (CD62E)
- Tests per kit:96 tests
- Primary antibody reactivity:Mouse
- Sample type:Citrate plasma, cell culture supernatant, serum
- Detection range:23,4 - 1500 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB201279-96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:8,55 pg/ml
- Format:Pre-coated
- Target protein:CD62E
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



