Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay Kit
ABCAAB109883-30
: Abcam
:
- Description:Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay kit
- Method:Sandwich
- Species reactivity:Other;Human;Mouse;Rat
- Form:Pre-coated
- Antigen name:Pyruvate dehydrogenase
- Storage temperature:4 °C
- Processing time:Multiple steps
- Regulatory status:RUO
- Tests per kit:30 tests
Pyruvate dehydrogenase (PDH) Protein Quantity Dipstick Assay Kit is an immunoassay kit for the detection of Pyruvate dehydrogenase (PDH) Protein Quantity in Mouse, Rat, Cow, Human in Cell culture extracts, Tissue samples.