Rat Interferon gamma ELISpot Pair
Catalog # ABCAAB281994-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat Interferon gamma ELISpot Pair
- Species reactivity:Rat
- Sample type:Suspension cells
- Form:Uncoated
- Storage temperature:+4 °C
- Processing time:Multiple steps
- Antigen symbol:Interferon gamma
- Regulatory status:RUO
Specifications
About this item
Rat Interferon gamma ELISpot Pair is a ELISpot Pair for the sensitive detection of Rat Interferon gamma production in a single cell suspension.