Order Entry
Germany
Orders LinkContactUsLinkComponent
Mouse Complement C5a ELISA Kit

Mouse Complement C5a ELISA Kit

Mouse Complement C5a ELISA kit, 96 tests
Catalog # ABCAAB193718-96
Supplier:   Abcam
Mouse Complement C5a ELISA Kit
Catalog # ABCAAB193718-96
Supplier:  Abcam
CAS Number:  
Mouse Complement C5a ELISA Kit
Mouse Complement C5a ELISA Kit

Mouse Complement C5a ELISA kit, 96 tests

CAS Number:  
Price
805,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Complement C5a ELISA kit ab193718 is used to quantify Mouse Complement C5a with a sensitivity of 6 pg/ml. Sandwich ELISA kit.

Specifications

Specifications

  • Description:
    Mouse Complement C5a ELISA kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB193718-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    1,5 pg/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Plasma, cell culture supernatant, serum
  • Target protein:
    C5
  • Detection range:
    1,5 - 400 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications