Rat Interferon gamma ELISPOT Kit (with un-coated plates)
: Abcam
Rat Interferon gamma ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Rat Interferon gamma production in a single cell suspension.
- Species reactivity:Rat
- Sample type:Suspension cells
- Antigen name:Interferon gamma
- Storage temperature:4 °C
- Processing time:Multiple steps
- Product type:Uncoated