Order Entry
Germany
Orders LinkContactUsLinkComponent
Rat TNF alpha ELISpot Pair

Rat TNF alpha ELISpot Pair

Rat TNF alpha ELISpot Pair
Catalog # ABCAAB281995-10X96
Supplier:   Abcam
Rat TNF alpha ELISpot Pair
Catalog # ABCAAB281995-10X96
Supplier:  Abcam
CAS Number:  
Rat TNF alpha ELISpot Pair
Rat TNF alpha ELISpot Pair

Rat TNF alpha ELISpot Pair

CAS Number:  
Price
1 310,00 €
Log in for availability
1 * 960 Tests
Product Details & Documents

Rat TNF alpha ELISpot Pair is a ELISpot Pair for the sensitive detection of Rat TNF alpha production in a single cell suspension.

Specifications

Specifications

  • Description:
    Rat TNF alpha ELISpot Pair
  • Cat. No.:
    ABCAAB281995-10X96
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Species reactivity:
    Rat
  • Storage temperature:
    4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    TNF alpha

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.