phynomic lite w Assembly Gloves, 60041 Series
Supplier: UVEX
Certificates
About this item
The uvex phynomic series is ideal for all types of work where a high level of feeling is required.
- Aqua-polymer impregnation on palm and fingertips
- Very high level of breathability with porous coating
- Outstanding tactile sensation when handling small parts
- Perfect fit with 3D Ergo technology
These safety gloves are like a 'second skin' and are especially light and flexible, without making hands immediately sweaty. The uvex phynomic lite is extremely lightweight, with a composition that makes it suited to particularly delicate tasks.
Specifications
- Catalog No:
- UVEX6004105
- UVEX6004112
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020/EN 388:2016 + A1:2018
- EN ISO 21420:2020/EN 388:2016 + A1:2018