Orders LinkContactUsLinkComponent
PCR tubes, Multiply®
PCR tubes, Multiply®
Marketsource
Supplier:   SARSTEDT
PCR tubes, Multiply®
Supplier:  SARSTEDT
Marketsource
Certificates
PCR tubes, Multiply®
13 Options Available Below
Product Details & Documents
PP, translucent
  • Thin, smooth walls ensure rapid and even heat transfer to the sample
  • Anti-contamination shield prevents accidental contact with the inner surface of the lid, preventing potential contamination
  • Frosted flat lid with large writing area
Developed due to the limitations of traditional reaction tubes they offer many desirable benefits. Enhanced design features ensure work is risk-free from contamination and non-tiring compared to the more traditional tubes. 0,2 and 0,5 ml thin-walled Multiply® PCR cups are moulded with an integral anti-contamination shield at the front of the lid.
Product Family Options
Single tubes
Product Information
  • Colour
    Capacity
    Availability
    Price
  • SARS72.737.002
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 109,00 €
    109,00 €
    1 * 2.000 ST
       109,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.735.002
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    Natural
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 73,80 €
    73,80 €
    1 * 2.000 ST
       73,80 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.735.992
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    Assorted
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 124,00 €
    124,00 €
    1 * 3.000 ST
       124,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.737.992
    PCR tubes, singles tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    PCR tubes, singles tubes, Multiply®-Pro with integral anti-contamination shield at the lid front
    Assorted
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 170,00 €
    170,00 €
    1 * 3.000 ST
       170,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.735.100
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front, Biosphere®, sterile
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front, Biosphere®, sterile
    Natural
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 52,40 €
    52,40 €
    1 * 1.000 ST
       52,40 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.737
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front, Biosphere®, sterile
    PCR tubes, single tubes, Multiply®-Pro with integral anti-contamination shield at the lid front, Biosphere®, sterile
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 117,00 €
    117,00 €
    1 * 2.000 ST
       117,00 €
    Specifications:
    Marketsource
    Marketsource
Strip of 4 tubes
Product Information
  • Colour
    Capacity
    Availability
    Price
  • SARS72.990
    PCR strip of 4 tubes, Biosphere® Multiply® -μStripPro, sterile, with attached lids
    PCR strip of 4 tubes, Biosphere® Multiply® -μStripPro, sterile, with attached lids
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 194,00 €
    194,00 €
    1 * 400 ST
       194,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.990.002
    PCR strip of 4 tubes, Multiply® -μStripPro with attached lids
    PCR strip of 4 tubes, Multiply® -μStripPro with attached lids
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 256,00 €
    256,00 €
    1 * 480 ST
       256,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.990.992
    PCR strip of 4 tubes, Multiply® -μStripPro with attached lids
    PCR strip of 4 tubes, Multiply® -μStripPro with attached lids
    Assorted
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 202,00 €
    202,00 €
    1 * 480 ST
       202,00 €
    Specifications:
    Marketsource
    Marketsource
Strip of 8 tubes
Product Information
  • Colour
    Capacity
    Availability
    Price
  • SARS72.985.002
    PCR strip of 8 tubes, Multiply® -μStripPro, without lids
    PCR strip of 8 tubes, Multiply® -μStripPro, without lids
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 318,00 €
    318,00 €
    1 * 480 ST
       318,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.991.002
    PCR strip of 8 tubes, Multiply® -μStripPro, with attached lids
    PCR strip of 8 tubes, Multiply® -μStripPro, with attached lids
    Natural
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 426,00 €
    426,00 €
    1 * 480 ST
       426,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.985.992
    PCR strip of 8 tubes, Multiply® -μStripPro, without lids
    PCR strip of 8 tubes, Multiply® -μStripPro, without lids
    Assorted
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 296,00 €
    296,00 €
    1 * 480 ST
       296,00 €
    Specifications:
    Marketsource
    Marketsource
  • SARS72.991.992
    PCR strip of 8 tubes, Multiply® -μStripPro with attached lids
    PCR strip of 8 tubes, Multiply® -μStripPro with attached lids
    Assorted
    0,2 ml
    Log in for availability
    Log in for availability
    1 * 480 ST
    1 * 480 ST - 367,00 €
    367,00 €
    1 * 480 ST
       367,00 €
    Specifications:
    Marketsource
    Marketsource