Cryoboxes
Marketsource
Hersteller: SARSTEDT
Jump to:
About this item
Cryoboxes, made of high quality durable polycarbonate.
- Numerical coding for each tubes enables quick sample identification
- Optically clear lid and coloured base with large apertures for swift ventilation
- Autoclavable
For storage of tubes at deep freeze range to −196 °C.
4 Options Available Below
Product Details & Documents
Cryoboxes, made of high quality durable polycarbonate.
- Numerical coding for each tubes enables quick sample identification
- Optically clear lid and coloured base with large apertures for swift ventilation
- Autoclavable
For storage of tubes at deep freeze range to −196 °C.
Product Family Options
Product Information
- CompartmentsForColourW×D×H ext.VerfügbarkeitPreis
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource