Order Entry
Germany
ContactUsLinkComponent
Petri dish, with 2 compartments
Petri dish, with 2 compartments
Catalog # SARS82.1195
Supplier:  SARSTEDT
Petri dish, with 2 compartments
Catalog # SARS82.1195
Supplier:  SARSTEDT

Specifications

  • Ø:
    92 mm
  • Packed:
    20/bag
  • Version:
    With 2 compartments, with vents
  • Height:
    16 mm
  • Pk:
    480

Specifications

About this item