Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Culture Dishes
Petri Dishes
Petri dishes, square
Petri dishes, square
Petri dishes, square
Catalog # SARS82.9923.422
Supplier:
SARSTEDT
Expand image
Petri dishes, square
Catalog # SARS82.9923.422
Supplier:
SARSTEDT
Specifications
Packed:
4/bag
Sterility:
Sterile
Version:
100×100 mm, without compartments
Height:
20 mm
Pk:
160
Specifications
About this item
Product Details
Product Details