Stirrer blades, PTFE, Quickfit®
Supplier: SciLabware
Certificates
About this item
Stirrer blades made of PTFE, and they can be pivoted on the glass stirrer shaft to allow passage through small apertures.
- Easily detachable for cleaning and replacement purposes
- Resistance to chemical attack
Suitable for sweeping close to the vessel walls as they will not cause scratching.