Cleanroom goggles, Coverall Autoclave
Catalog # BOLECOVACLAVE
Supplier: Bollé Safety
Specifications
- Type:Anti-scratch, anti-fog, chemical-resistant
- Seq:02
- Lenses:Clear PC
- Pk:1
Specifications
About this item
These cleanroom Coverall goggles with interchangeable ventilation ensure protection and comfort. It is possible to wear prescription spectacles under them.
- PLATINUM coated lens: Coating on both sides ensures high scratch-resistance (1,4 cd/m²), high resistance to the most aggressive chemicals and slows the appearance of fogging
- Adjustable strap
- Upper indirect and interchangeable ventilation
- Ultra-comfort autoclavable goggles
Weight: 54 g
Certifications: EN 166