Order Entry
Germany
ContactUsLinkComponent
Decabromodiphenyl ether (BDE No. 209)
Decabromodiphenyl ether (BDE No. 209)
Catalog # EHERC15898209
Supplier:  EHRENSTORFER
CAS Number:  
Decabromodiphenyl ether (BDE No. 209)
Catalog # EHERC15898209
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
EINECS:214-604-9
CAS Number:1163-19-5
Formula:C₁₂Br₁₀O
MDL Number:MFCD00000059