Chromone-2-carboxylic acid
Catalog # MOLE13834204-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:190.15 g/mol
EINECS:225-583-0
CAS Number:4940-39-0
Formula:C₁₀H₆O₄
MDL Number:MFCD00006838