Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Ethoxy-4-methoxybenzaldehyde
3-Ethoxy-4-methoxybenzaldehyde
3-Ethoxy-4-methoxybenzaldehyde
3-Ethoxy-4-methoxybenzaldehyde
Supplier:
CHEMPUR
Certificates
Expand image
3-Ethoxy-4-methoxybenzaldehyde
Supplier:
CHEMPUR
About this item
CAS Number:
1131-52-8
:
Formula:
C₁₀H₁₂O₃
MDL Number:
MFCD00010128
:
180.20 g/mol