Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluorene-[D10]
Fluorene-[D₁₀] (98% D), Sigma-Aldrich®
Fluorene-[D₁₀] (98% D), Sigma-Aldrich®
Fluorene-[D₁₀] (98% D), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Fluorene-[D₁₀] (98% D), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
81103-79-9
:
Formula:
C₁₃D₁₀
MDL Number:
MFCD00144313
ADR:
9,III
:
M-Clarity™ quality level = MQ200
Storage Temperature:
Ambient
UN:
3077