Order Entry
Germany
ContactUsLinkComponent
Fluorene-[D₁₀]
Fluorene-[D₁₀]
Catalog # EHERC20800200
Supplier:  EHRENSTORFER
CAS Number:  
Fluorene-[D₁₀]
Catalog # EHERC20800200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3077
ADR:9,III
CAS Number:81103-79-9
Formula:C₁₃D₁₀
MDL Number:MFCD00144313