Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluorene-[D10]
Fluorene-[D₁₀] (98% D)
Fluorene-[D₁₀] (98% D)
Fluorene-[D₁₀] (98% D)
Supplier:
LGC Standards PROMOCHEM
Certificates
Expand image
Fluorene-[D₁₀] (98% D)
Supplier:
LGC Standards PROMOCHEM
About this item
CAS Number:
81103-79-9
:
Formula:
C₁₃D₁₀
MDL Number:
MFCD00144313
Storage Temperature:
Ambient
UN:
3077
ADR:
9,III