Order Entry
Germany
Orders LinkContactUsLinkComponent
Thioisonicotinamide

Thioisonicotinamide

Glass bottle for solids, 100 g
Catalog # MOLEM23097848
Marketsource
Supplier:   Molekula
Thioisonicotinamide
Catalog # MOLEM23097848
Supplier:  Molekula
Marketsource
CAS Number:  2196-13-6
Thioisonicotinamide

Glass bottle for solids, 100 g

CAS Number:  2196-13-6
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • MDL Number:MFCD00006437
  • Molecular Weight:138.19 g/mol
  • EINECS:218-592-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.