Order Entry
Germany
Orders LinkContactUsLinkComponent
Thioisonicotinamide

Thioisonicotinamide

Glass bottle for solids, 10 g
Catalog # FLUO021346-10G
Marketsource
Supplier:   FLUOROCHEM
Thioisonicotinamide
Catalog # FLUO021346-10G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  2196-13-6
Thioisonicotinamide

Glass bottle for solids, 10 g

CAS Number:  2196-13-6
Price
118,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • MDL Number:MFCD00006437
  • Molecular Weight:138.19 g/mol
  • EINECS:218-592-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications