Thioisonicotinamide
Supplier: Cayman Chemical
Jump to:
About this item
Synonyms: Pyridine-4-carbothioamide
- CAS Number:2196-13-6
- Formula:C₆H₆N₂S
- MDL Number:MFCD00006437
- Molecular Weight:138.19 g/mol
- EINECS:218-592-6
4 Options Available Below
Product Details & Documents
Synonyms: Pyridine-4-carbothioamide
- CAS Number:2196-13-6
- Formula:C₆H₆N₂S
- MDL Number:MFCD00006437
- Molecular Weight:138.19 g/mol
- EINECS:218-592-6



