Thioisonicotinamide 97%
Glass bottle, 25 g
Supplier: Thermo Fisher Scientific
Thioisonicotinamide 97%
Catalog # ACRO138770250
Supplier: Thermo Fisher Scientific
CAS Number: 2196-13-6
Jump to:
Product Details & Documents
Synonyms: Pyridine-4-carbothioamide
- CAS Number:2196-13-6
- Formula:C₆H₆N₂S
- MDL Number:MFCD00006437
- Molecular Weight:138.19 g/mol
- Melting Pt:198…202 °C
- Storage Temperature:Ambient
- EINECS:218-592-6
Documents
ACRO13877
SDS
Specifications
Specification Test Results
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:193 to 201 °C
- HPLC:≥96.0 %
Specifications
- Pack type:Glass bottle
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



