Nicotinoyl chloride hydrochloride
Catalog # MOLE15331400-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:178.02 g/mol
EINECS:243-648-1
UN:3261
ADR:8,II
CAS Number:20260-53-1
Formula:C₆H₅Cl₂NO
MDL Number:MFCD00012820