2-(4-Fluorophenoxy)benzaldehyde
Glass bottle for solids, 1 g
Catalog # FLUO010111-1G
Marketsource
Supplier: FLUOROCHEM
2-(4-Fluorophenoxy)benzaldehyde
Catalog # FLUO010111-1G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 320423-61-8
Jump to:
Product Details & Documents
- CAS Number:320423-61-8
- Formula:C₁₃H₉FO₂
- MDL Number:MFCD01568770
- Molecular Weight:216.21 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



