2-(4-Fluorophenoxy)benzaldehyde ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 320423-61-8 Molecular Formula: C13H9FO2 Formula Weight: 216.21 MDL No.: MFCD01568770
- CAS Number:320423-61-8
- Formula:C₁₃H₉FO₂
- MDL Number:MFCD01568770
- Molecular Weight:216.21 g/mol
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 320423-61-8 Molecular Formula: C13H9FO2 Formula Weight: 216.21 MDL No.: MFCD01568770
- CAS Number:320423-61-8
- Formula:C₁₃H₉FO₂
- MDL Number:MFCD01568770
- Molecular Weight:216.21 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



