3-(1,4-Cyclohexadien-1-yl)-1-propanol ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87151-66-4
:
Formula:C₉H₁₄O
MDL Number:MFCD12407081
:138.21 g/mol
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Storage Temperature:Freezer