0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥98%
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥98%
:
Thermo Fisher Scientific
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)aniline ≥98%
:
Thermo Fisher Scientific
:
651-83-2
:
:
C₇H₂F₇N
:
MFCD00091518
:
233.09 g/mol
:
185…187 °C
:
Ambient
:
2810
:
6.1,III
:
GHS07
msds_ALFAB24326
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years