Perfluorohexane, Sigma-Aldrich®
SIAL379247-50ML
: Merck
:
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
:
:338.04 g/mol
:56 °C (1013 hPa)
:–4 °C
:1.68 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:206-585-0
:GHS07
:355-42-0
:C₆F₁₄
:MFCD00000437