0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Potassium permanganate
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
:
Bernd Kraft
Potassium permanganate 0.02 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur.
:
Bernd Kraft
:
7722-64-7
:
:
KMnO₄
:
MFCD00011364
:
158.03 g/mol
:
Ambient
:
231-760-3
:
3082
:
9,III
:
GHS09