1,1,1,3,5,5,5-Heptamethyltrisiloxane
Catalog # ABCRSIB1844.0.0010
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
About this item
:
:222.51 g/mol
EINECS:217-496-1
CAS Number:1873-88-7
Formula:C₇H₂₂O₂Si₃
MDL Number:MFCD00048000