2-Chloro-5-(trifluoromethyl)aniline
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:121-50-6
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00007673
Boiling Pt:208 °C (1013 hPa)
Melting Pt:10 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-475-7
UN:2810
ADR:6.1,III
:195.57 g/mol