Microtome blades, disposable, FEATHER®
Supplier: PFM Medical GmbH
Certificates
About this item
Made of stainless steel. FEATHER® disposable microtome blades are used for paraffin and cryostat sectioning for the whole range of individual applications in routine practice, research and industry. The ultra-precision edge of these disposable microtome blades is specially coated for superior cutting performance.
- Blade angle: 35° and 22° (A22 and S22)
- Very high sectioning quality from the first cut
- Long service life
- Very short trimming time
- Edge finish: Resin coating, platinum spattering