TSK-Gel® alpha and superaw columns for gel filtration
Supplier: TOSOH Bioscience
Certificates
About this item
These columns are packed with a hydrophilic, highly crosslinked polymer used for a wide range of solvents. Suitable for the separations of synthetic polymers, oligomers, additives and detergents, saccharides, nucleic acids and peptides.
- Unique hydrophilic polymethacrylate resin with rigid spherical beads
- Minimal swelling characteristics from 100% water to 100% non-polar solvents
- Excellent mechanical and chemical stability
- High resolution power and short analysis time