Chloro(1,5-cyclooctadiene)iridium(I) dimer
Catalog # COBBOS-7525-1G
Supplier: COMBI-BLOCKS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:671.70 g/mol
EINECS:235-170-7
CAS Number:12112-67-3
Formula:C₁₆H₂₄Cl₂Ir₂
MDL Number:MFCD00012414