
Chloro(1,5-cyclooctadiene)iridium(I) dimer (57.2% Ir)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12112-67-3
:
Formula:C₁₆H₂₄Cl₂Ir₂
MDL Number:MFCD00012414
:671.70 g/mol
Melting Pt:190 °C (dec.)
EINECS:235-170-7
:GHS07
Storage Temperature:Ambient