Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chloro(1,5-cyclooctadiene)iridium (I) dimer
Chloro(1,5-cyclooctadiene)iridium(I) dimer
Chloro(1,5-cyclooctadiene)iridium(I) dimer
:
Thermo Fisher Scientific
Chloro(1,5-cyclooctadiene)iridium(I) dimer
:
Thermo Fisher Scientific
:
12112-67-3
:
:
C₁₆H₂₄Cl₂Ir₂
:
MFCD00012414
:
671.70 g/mol
:
190 °C (dec.)
:
235-170-7
:
GHS07
:
Ambient
Appearance:
Orange to red Powder
Infrared spectrum:
Conforms
Iridium (Ir):
≥56.4 %
ACRO36396