Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Benzo[a]pyrene-[D12] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Benzo[a]pyrene-[D12] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Benzo[a]pyrene-[D12] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Supplier:
EHRENSTORFER
Certificates
Expand image
Benzo[a]pyrene-[D12] 10 µg/ml in cyclohexane, Dr Ehrenstorfer
Supplier:
EHRENSTORFER
About this item
Specifications
CAS No.:
63466-71-7
Formula:
C₂₀D₁₂
Description:
Benzo[a]pyrene-[D12]
Specifications