1-(Trifluoromethyl)cyclopropanecarboxylic acid
Catalog # FLUO009450-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:154.09 g/mol
UN:3261
ADR:8,III
CAS Number:277756-46-4
Formula:C₅H₅F₃O₂
MDL Number:MFCD03093070