(±)-Dihydrolipoic acid, Millipore®
Catalog # 437694-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
CAS Number:7516-48-5
Formula:C₈H₁₆O₂S₂
MDL Number:MFCD00166974