Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(1H-Imidazol-1-yl)phenol
4-(1H-Imidazol-1-yl)phenol, Sigma-Aldrich®
4-(1H-Imidazol-1-yl)phenol, Sigma-Aldrich®
4-(1H-Imidazol-1-yl)phenol, Sigma-Aldrich®
SIAL183725-1G
:
Merck
:
4-(1H-Imidazol-1-yl)phenol, Sigma-Aldrich®
SIAL183725-1G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
160.18 g/mol
:
M-Clarity™ quality level = MQ100
:
233-121-4
:
10041-02-8
:
C₉H₈N₂O
:
MFCD00005281