Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2-Bromophenyl)pyrrole
1-(2-Bromophenyl)pyrrole
1-(2-Bromophenyl)pyrrole
1-(2-Bromophenyl)pyrrole
Supplier:
BIONET RESEARCH
Certificates
Expand image
1-(2-Bromophenyl)pyrrole
Supplier:
BIONET RESEARCH
About this item
CAS Number:
69907-27-3
:
Formula:
C₁₀H₈BrN
MDL Number:
MFCD02665226
:
222.0800 g/mol