Order Entry
Germany
ContactUsLinkComponent
Butamben (butyl-4-aminobenzoate)
Butamben (butyl-4-aminobenzoate)
Catalog # MOLE55959005-5G
Supplier:  Molekula
CAS Number:  
Butamben (butyl-4-aminobenzoate)
Catalog # MOLE55959005-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :193.25 g/mol
EINECS:202-317-1
CAS Number:94-25-7
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00017112