Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Butamben (Butyl 4-aminobenzoate)
Butamben (butyl-4-aminobenzoate) 99%
Butamben (butyl-4-aminobenzoate) 99%
:
Thermo Fisher Scientific
Butamben (butyl-4-aminobenzoate) 99%
:
Thermo Fisher Scientific
:
94-25-7
:
:
C₁₁H₁₅NO₂
:
MFCD00017112
:
193.25 g/mol
:
14,1512
:
174 °C (8 mmHg)
:
56…59 °C
:
Ambient
:
202-317-1
:
GHS07
msds_ALFAH54690
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years