Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Turbidity Standards
Turbidity standard, 4000 FNU
Turbidity standard, 4000 FNU
Turbidity standard, 4000 FNU
Catalog # HACHLCW813
Supplier:
Hach
CAS Number:
Certificates
Expand image
Turbidity standard, 4000 FNU
Catalog # HACHLCW813
Supplier:
Hach
CAS Number:
Certificates
Specifications
Pack type:
Glass bottle
Pk:
100 mL
Turbidity:
4000 FNU
Specifications
About this item
Product Details
Documentation
Product Details
Documentation