Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate) 10% in aqueous solution
Tween® 20 (Polysorbate) 10% in aqueous solution
Tween® 20 (Polysorbate) 10% in aqueous solution
Supplier:
Bernd Kraft
Certificates
Expand image
Tween® 20 (Polysorbate) 10% in aqueous solution
Supplier:
Bernd Kraft
About this item
CAS Number:
9005-64-5
:
Formula:
C₅₈H₁₁₄O₂₆
MDL Number:
MFCD00165986
:
1227.53 g/mol
Storage Temperature:
Ambient
EINECS:
500-018-3