Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Tween® 20 (Polysorbate)

Supplier:   Bernd Kraft
Tween® 20 (Polysorbate)
Supplier:  Bernd Kraft
Certificates
Tween® 20 (Polysorbate)

3 Options Available Below

Product Details & Documents
Synonyms:  Polyoxyethylene sorbitan monolaurate
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Molecular Weight:1227.53 g/mol
  • Density:1.1 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 64,10 €
    64,10 €
    1 * 500 mL
       64,10 €

  • Plastic bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 229,00 €
    229,00 €
    1 * 2.500 mL
       229,00 €

  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 111,00 €
    111,00 €
    1 * 1 L
       111,00 €