VWR® Autoclavable Bags, 'Sterilbag' and 'Sterilbag indicator'
Supplier: VWR Collection
Certificates
About this item
These bags are ideal for autoclaving polystyrene containers and other items contaminated by pathogenic agents (petri dishes, microtitre plates, pipettes, bottles, tubes, etc). They can be used for realising and preserving sterile conditions. Made of special plastic, red coloured.
- Autoclavable up to 134 °C
- Available with or without sterilisation indicator strip
- Red coloured
- Instructions printed on the bag