Order Entry
Germany
ContactUsLinkComponent
Fluoroplastic (PTFE) spray
Fluoroplastic (PTFE) spray
  BOHLH958-04
 :  Bohlender
undefined
Fluoroplastic (PTFE) spray
  BOHLH958-04
 :  Bohlender

 

  • Description:
    Fluroplastic spray, PTFE, 400 ml
  • Pk:
    1

 

 

Release and anti-blocking agent as well as lubricant for laboratory. Excellent gliding and separating effects.

  • Dry and greaseless
  • CFC- and silicone-free
  • Heat resistant up to +260°C
  • Non-adhesive and dirt-repellent
  • Physiologically safe